Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007734.2 | NP_031760.2 | 52 | 5389 | ||
GenBank mRNA | AF169387.1 | - | 52 | 5334 | ||
AK033387.1 | - | 3 | 296 | |||
Z35166.1 | - | 5 | 696 |