Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243008.1 | NP_001229937.1 | 11 | 5374 | ||
NM_001243009.1 | NP_001229938.1 | 8 | 3553 | |||
GenBank mRNA | AF064749.1 | - | 8 | 3360 | ||
AK076391.1 | - | 3 | 1356 | |||
Z18279.1 | - | 1 | 59 |