Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017429.2 | NP_001017429.1 | 2 | 242 | ||
GenBank mRNA | AK146388.1 | - | 2 | 199 | ||
AK148493.1 | - | 2 | 210 | |||
AK152968.1 | - | 2 | 203 | |||
AK158051.1 | - | 2 | 220 | |||
AK160687.1 | - | 1 | 2235 | |||
BC048668.1 | - | 2 | 242 | 6773943 | ||
BG792513.1 | - | 2 | 174 |