Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007748.3 | NP_031774.1 | 3 | 392 | ||
GenBank mRNA | AK002402.1 | - | 3 | 394 | ||
AK019318.1 | - | 3 | 397 | |||
AK166496.1 | - | 3 | 394 | |||
BC003807.1 | - | 3 | 392 | 3487598 | ||
BC052816.1 | - | 3 | 398 | 6439987 | ||
L06465.1 | - | 3 | 391 | |||
U08440.1 | - | 3 | 381 |