Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004998.3 | NP_004989.2 | 10 | 1305 | ||
GenBank mRNA | AB290177.1 | - | 10 | 1288 | ||
AK291688.1 | - | 10 | 1302 | |||
BC098392.1 | - | 10 | 1304 | 30527536 | ||
U14391.1 | - | 10 | 1308 |