Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007754.2 | NP_031780.2 | 12 | 2728 | ||
GenBank mRNA | AK032306.1 | - | 12 | 2220 | ||
AK134907.1 | - | 14 | 2663 | |||
AK137737.1 | - | 12 | 2707 | |||
D85391.1 | - | 12 | 2716 |