Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007754.2 | NP_031780.2 | 13 | 3091 | ||
GenBank mRNA | AK032306.1 | - | 13 | 2583 | ||
AK137737.1 | - | 13 | 3070 | |||
D85391.1 | - | 13 | 3079 |