Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004455.2 | NP_001004455.2 | 5 | 1666 | ||
NM_001161807.1 | NP_001155279.1 | 5 | 1706 | |||
NM_138686.3 | NP_619627.3 | 5 | 1737 | |||
GenBank mRNA | AF390548.1 | - | 5 | 1755 | ||
AK013490.1 | - | 5 | 898 | |||
CF613278.1 | - | 1 | 385 |