Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004455.2 | NP_001004455.2 | 5 | 1559 | ||
NM_001161807.1 | NP_001155279.1 | 5 | 1599 | |||
NM_138686.3 | NP_619627.3 | 5 | 1630 | |||
GenBank mRNA | AF390548.1 | - | 5 | 1648 | ||
AK013490.1 | - | 5 | 791 | |||
BE996554.1 | - | 5 | 533 |