Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cplx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007756.3 | NP_031782.3 | 4 | 1292 | ||
GenBank mRNA | AK134820.1 | - | 4 | 1235 | ||
AK158166.1 | - | 4 | 1235 | |||
AK159026.1 | - | 4 | 1251 | |||
BC014803.1 | - | 4 | 1197 | 4527117 |