Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009949.2 | NP_034079.2 | 4 | 825 | ||
GenBank mRNA | AK144441.1 | - | 4 | 825 | ||
AK150163.1 | - | 4 | 797 | |||
AK169038.1 | - | 4 | 722 | |||
BC138514.1 | - | 4 | 760 | 8861534 | ||
BC145859.1 | - | 4 | 760 | 40131162 | ||
U01163.1 | - | 4 | 770 |