Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007758.2 | NP_031784.1 | 15, 16 | 3013 | ||
GenBank mRNA | AK003072.2 | - | 15, 16 | 3013 | ||
AK007487.1 | - | 1, 2 | 167 | |||
BC109344.1 | - | 15, 16 | 2946 | 40054528 | ||
M29281.1 | - | 10, 11 | 2017 |