Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007759.2 | NP_031785.1 | 2 | 338 | ||
GenBank mRNA | BC018397.1 | - | 2 | 296 | 3588770 | |
M35523.1 | - | 2 | 305 |