Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007760.3 | NP_031786.2 | 9 | 1563 | ||
GenBank mRNA | AK028666.1 | - | 3 | 208 | ||
AK041431.1 | - | 9 | 1338 | |||
AK132179.1 | - | 10 | 1390 | |||
AK170740.1 | - | 9 | 1349 | |||
BC006668.1 | - | 9 | 1310 | 3499169 | ||
X85983.1 | - | 9 | 1281 |