Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198839.1 | NP_001185768.1 | 5 | 1915 | ||
NM_009954.3 | NP_034084.2 | 5 | 1978 | |||
GenBank mRNA | AK079554.1 | - | 4 | 1941 | ||
AK090315.1 | - | 5 | 1989 | |||
AK145863.1 | - | 5 | 1979 | |||
AK159168.1 | - | 5 | 1882 | |||
AK159297.1 | - | 5 | 1883 | |||
AK166248.1 | - | 5 | 1853 | |||
AK167352.1 | - | 5 | 1916 | |||
AK167504.1 | - | 5 | 1916 | |||
AK167516.1 | - | 5 | 1980 | |||
AK167717.1 | - | 5 | 1979 | |||
AK177203.1 | - | 1 | 87 | |||
BC057578.1 | - | 5 | 1938 | 6401186 | ||
U28151.1 | - | 5 | 1889 | |||
U48853.1 | - | 5 | 1873 |