Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009955.3 | NP_034085.2 | 10 | 1244 | ||
GenBank mRNA | BC062955.1 | - | 10 | 1243 | 30535984 | |
X87242.1 | - | 10 | 1141 | |||
Y10339.1 | - | 10 | 1034 |