Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021351.1 | NP_067326.1 | 4 | 397 | ||
GenBank mRNA | AJ272228.2 | - | 4 | 397 | ||
AK132591.1 | - | 5 | 414 | |||
AK161601.1 | - | 3 | 361 | |||
AK162018.1 | - | 5 | 411 | |||
BC056444.1 | - | 4 | 387 | 6824277 | ||
BC058703.1 | - | 4 | 397 | 6824293 |