Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159650.1 | NP_001153122.1 | 5 | 411 | ||
NM_021352.3 | NP_067327.1 | 5 | 481 | |||
GenBank mRNA | AJ272229.2 | - | 4 | 465 | ||
AK053552.1 | - | 3 | 795 | |||
BC031442.1 | - | 4 | 472 | 4501690 | ||
BC058522.1 | - | 4 | 390 | 6824229 | ||
CJ046471.1 | - | 5 | 482 |