Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016669.1 | NP_057878.1 | 3 | 399 | ||
GenBank mRNA | AF039391.1 | - | 3 | 399 | ||
AK138422.1 | - | 3 | 370 | |||
AK143116.1 | - | 3 | 389 | |||
AK163781.1 | - | 3 | 370 | |||
AK163801.1 | - | 3 | 396 | |||
BC045159.1 | - | 3 | 360 | 6492374 |