Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037859.2 | NP_001032948.2 | 4 | 865 | ||
GenBank mRNA | AK004947.1 | - | 3 | 675 | ||
AK041299.1 | - | 3 | 653 | |||
AK076215.1 | - | 3 | 673 | |||
AK079247.1 | - | 3 | 682 | |||
AK080326.1 | - | 3 | 662 | |||
AK089498.1 | - | 3 | 655 | |||
AK143545.1 | - | 3 | 666 | |||
AK146069.1 | - | 4 | 866 | |||
AK154128.1 | - | 3 | 680 | |||
AK154556.1 | - | 3 | 676 | |||
AK154653.1 | - | 3 | 676 | |||
AK154699.1 | - | 3 | 676 | |||
AK155488.1 | - | 3 | 685 | |||
AK155565.1 | - | 3 | 695 | |||
AK155576.1 | - | 682 | ||||
AK155640.1 | - | 3 | 680 | |||
AK155654.1 | - | 3 | 680 | |||
AK155858.1 | - | 3 | 683 | |||
AK156550.1 | - | 3 | 700 | |||
AK171241.1 | - | 3 | 715 | |||
AK171386.1 | - | 3 | 680 | |||
AK171543.1 | - | 3 | 655 | |||
BC036343.1 | - | 3 | 652 | 4948135 | ||
BC043054.1 | - | 3 | 656 | 6418718 | ||
BC050024.1 | - | 3 | 722 | |||
BC066863.1 | - | 4 | 774 | 30436119 | ||
X06368.1 | - | 3 | 658 | |||
X68932.1 | - | 3 | 658 |