Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037859.2 | NP_001032948.2 | 20, 21 | 2928 | ||
GenBank mRNA | AK004947.1 | - | 19, 20 | 2738 | ||
AK076215.1 | - | 19, 20 | 2737 | |||
AK079247.1 | - | 19, 20 | 2745 | |||
AK080326.1 | - | 19, 20 | 2726 | |||
AK143545.1 | - | 19, 20 | 2729 | |||
AK146069.1 | - | 20, 21 | 2929 | |||
AK154128.1 | - | 19, 20 | 2743 | |||
AK154556.1 | - | 19, 20 | 2739 | |||
AK154653.1 | - | 19, 20 | 2739 | |||
AK154699.1 | - | 19, 20 | 2739 | |||
AK155488.1 | - | 19, 20 | 2749 | |||
AK155498.1 | - | 5, 6 | 505 | |||
AK155565.1 | - | 15, 16 | 2238 | |||
AK155576.1 | - | 2745 | ||||
AK155640.1 | - | 19, 20 | 2743 | |||
AK155654.1 | - | 19, 20 | 2743 | |||
AK155858.1 | - | 19, 20 | 2746 | |||
AK156550.1 | - | 19, 20 | 2763 | |||
AK171386.1 | - | 19, 20 | 2743 | |||
BC036343.1 | - | 19, 20 | 2715 | 4948135 | ||
BC043054.1 | - | 19, 20 | 2719 | 6418718 | ||
BC050024.1 | - | 19, 20 | 2826 | |||
BC066863.1 | - | 20, 21 | 2837 | 30436119 | ||
X06368.1 | - | 19, 20 | 2718 | |||
X68932.1 | - | 19, 20 | 2628 |