Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037859.2 | NP_001032948.2 | 15 | 2295 | ||
GenBank mRNA | AK004947.1 | - | 14 | 2105 | ||
AK041299.1 | - | 14 | 2083 | |||
AK076215.1 | - | 14 | 2104 | |||
AK079247.1 | - | 14 | 2112 | |||
AK080326.1 | - | 14 | 2093 | |||
AK143545.1 | - | 14 | 2096 | |||
AK146069.1 | - | 15 | 2296 | |||
AK154128.1 | - | 14 | 2110 | |||
AK154556.1 | - | 14 | 2106 | |||
AK154653.1 | - | 14 | 2106 | |||
AK154699.1 | - | 14 | 2106 | |||
AK155488.1 | - | 14 | 2116 | |||
AK155576.1 | - | 2112 | ||||
AK155640.1 | - | 14 | 2110 | |||
AK155654.1 | - | 14 | 2110 | |||
AK155858.1 | - | 14 | 2113 | |||
AK156550.1 | - | 14 | 2130 | |||
AK171386.1 | - | 14 | 2110 | |||
BC036343.1 | - | 14 | 2082 | 4948135 | ||
BC043054.1 | - | 14 | 2086 | 6418718 | ||
BC050024.1 | - | 14 | 2193 | |||
BC066863.1 | - | 15 | 2204 | 30436119 | ||
X68932.1 | - | 14 | 1998 |