Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301334.1 | NP_001288263.1 | 8, 9 | 287 | ||
NM_001301335.1 | NP_001288264.1 | 7, 8 | 254 | |||
NM_009973.3 | NP_034103.2 | 8, 9 | 287 | |||
GenBank mRNA | AK021337.1 | - | 8, 9 | 283 | ||
AK052780.1 | - | 8, 9 | 285 | |||
AK085744.1 | - | 8, 9 | 287 | |||
AK085747.1 | - | 8, 9 | 285 | |||
AK164763.1 | - | 8, 9 | 285 | |||
AK164767.1 | - | 8, 9 | 287 | |||
AK164768.1 | - | 8, 9 | 285 | |||
AK164795.1 | - | 8, 9 | 283 | |||
BC002084.1 | - | 8, 9 | 268 | 3489886 | ||
J00379.1 | - | 7, 8 | 253 | |||
V00740.1 | - | 7, 8 | 253 |