Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301334.1 | NP_001288263.1 | 10 | 343 | ||
NM_001301335.1 | NP_001288264.1 | 9 | 325 | |||
NM_009973.3 | NP_034103.2 | 10 | 358 | |||
GenBank mRNA | AK021337.1 | - | 10 | 354 | ||
AK052780.1 | - | 10 | 356 | |||
AK085744.1 | - | 10 | 358 | |||
AK085747.1 | - | 10 | 356 | |||
AK142683.1 | - | 516 | ||||
AK164763.1 | - | 10 | 341 | |||
AK164767.1 | - | 10 | 358 | |||
AK164768.1 | - | 10 | 356 | |||
AK164795.1 | - | 10 | 354 | |||
BC002084.1 | - | 10 | 339 | 3489886 | ||
J00379.1 | - | 9 | 324 | |||
V00740.1 | - | 9 | 324 |