Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258040.2 | NP_001244969.1 | 10 | 1582 | ||
NM_001258041.2 | NP_001244970.1 | 11 | 1642 | |||
NM_001258042.2 | NP_001244971.1 | 10 | 1522 | |||
NM_001289092.1 | NP_001276021.1 | 9 | 1480 | |||
NM_001289093.1 | NP_001276022.1 | 8 | 1360 | |||
NM_001289094.1 | NP_001276023.1 | 11 | 1615 | |||
NM_002109.5 | NP_002100.2 | 11 | 1702 | |||
GenBank mRNA | AK000498.1 | - | 11 | 1382 | ||
AK094406.1 | - | 2 | 1155 | |||
AK124831.1 | - | 10 | 2892 | |||
AK225776.1 | - | 11 | 1292 | |||
AK293154.1 | - | 9 | 1094 | |||
AK295219.1 | - | 10 | 1178 | |||
AK302295.1 | - | 10 | 1196 | |||
AK303778.1 | - | 8 | 1170 | |||
AK307669.1 | - | 11 | 1295 | |||
AY995220.1 | - | 11 | 1279 | |||
BC011807.2 | - | 11 | 1309 | 4331277 | ||
BC080514.1 | - | 11 | 1307 | 2821065 | ||
X05345.1 | - | 11 | 1354 | |||
Z11518.1 | - | 11 | 1837 |