Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007786.4 | NP_031812.2 | 4 | 540 | ||
GenBank mRNA | AK021350.1 | - | 4 | 567 | ||
AK085710.1 | - | 4 | 567 | |||
AK142607.1 | - | 4 | 541 | |||
AK142673.1 | - | 4 | 541 | |||
AK164788.1 | - | 4 | 567 | |||
BC035443.1 | - | 4 | 550 | |||
BC049949.1 | - | 4 | 550 | 3963894 | ||
BC050269.1 | - | 4 | 545 | 3966924 | ||
M10114.1 | - | 4 | 525 |