Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007788.3 | NP_031814.2 | 12 | 1263 | ||
GenBank mRNA | AK146032.1 | - | 12 | 1222 | ||
AK158077.1 | - | 12 | 1193 | |||
BC026149.1 | - | 12 | 1167 | 4188101 | ||
BC060742.1 | - | 12 | 1218 | 4945773 | ||
BC089343.1 | - | 12 | 1141 | 30360017 | ||
U17112.1 | - | 11 | 1002 | |||
U51866.1 | - | 12 | 1205 |