Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297436.1 | NP_001284365.1 | 3, 4 | 981 | ||
NM_001523.3 | NP_001514.2 | 3, 4 | 984 | |||
GenBank mRNA | AY916556.1 | - | 2, 3 | 281 | ||
BC035837.1 | - | 3, 4 | 963 | 5589083 | ||
D84424.1 | - | 2, 3 | 968 | |||
U59269.1 | - | 3, 4 | 960 |