Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081249.1 | NP_001074718.1 | 5 | 1100 | ||
NM_001134474.1 | NP_001127946.1 | 5 | 1100 | |||
NM_001134475.1 | NP_001127947.1 | 5 | 1100 | |||
NM_019389.2 | NP_062262.2 | 5 | 1100 | |||
NM_172955.1 | NP_766543.1 | 5 | 1100 | |||
GenBank mRNA | AK014525.1 | - | 5 | 910 | ||
AK034871.1 | - | 5 | 907 | |||
AK040503.1 | - | 2 | 234 | |||
AK135324.1 | - | 5 | 1097 | |||
AK163927.1 | - | 5 | 957 | |||
AK165069.1 | - | 5 | 903 | |||
BC021652.1 | - | 5 | 940 | |||
BC096495.1 | - | 5 | 910 | |||
D16263.1 | - | 4 | 660 | |||
D28599.1 | - | 5 | 832 | |||
D32040.1 | - | 5 | 832 |