Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009977.3 | NP_034107.2 | 2 | 306 | ||
GenBank mRNA | AB015224.1 | - | 2 | 287 | ||
AF031825.1 | - | 2 | 235 | |||
AK004420.1 | - | 2 | 307 | |||
AK170583.1 | - | 2 | 253 | |||
BC117047.1 | - | 2 | 291 | 40126366 | ||
BC117049.1 | - | 2 | 291 | 40126368 |