Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009978.2 | NP_034108.1 | 3 | 435 | ||
GenBank mRNA | AF090691.1 | - | 3 | 424 | ||
AK018943.1 | - | 3 | 436 | |||
AK054409.1 | - | 3 | 418 | |||
AK161808.1 | - | 3 | 428 | |||
BC049753.1 | - | 3 | 440 | 6773192 | ||
S49926.1 | - | 3 | 236 |