Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009980.4 | NP_034110.1 | 3, 4 | 600 | ||
GenBank mRNA | AB033123.1 | - | 3, 4 | 465 | ||
AF016508.1 | - | 1, 2 | 72 | |||
AF059735.1 | - | 1, 2 | 51 | |||
AK036479.1 | - | 1 | 1647 | |||
AK146713.1 | - | 3, 4 | 485 | |||
AK147871.1 | - | 3, 4 | 483 | |||
AK178844.1 | - | 3, 4 | 274 |