Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007799.3 | NP_031825.2 | 8, 9 | 1286 | ||
GenBank mRNA | AK143581.1 | - | 7, 8 | 988 | ||
AK145875.1 | - | 8, 9 | 1143 | |||
AK157907.1 | - | 7, 8 | 1008 | |||
AK165271.1 | - | 7, 8 | 954 | |||
BC005432.1 | - | 8, 9 | 1286 | 3601519 | ||
CT010252.1 | - | 7, 8 | 922 | |||
X97399.1 | - | 7, 8 | 973 |