Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007803.5 | NP_031829.2 | 10, 11 | 1127 | ||
GenBank mRNA | AK146856.1 | - | 10, 11 | 934 | ||
AK148010.1 | - | 10, 11 | 930 | |||
AK168699.1 | - | 10, 11 | 928 | |||
BC117820.1 | - | 9, 10 | 829 | |||
U03184.1 | - | 10, 11 | 916 |