Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cux2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007804.2 | NP_031830.2 | 5 | 557 | ||
GenBank mRNA | AK006762.1 | - | 2 | 108 | ||
BC065113.1 | - | 5 | 557 | 30532644 | ||
U45665.1 | - | 5 | 378 |