Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007805.4 | NP_031831.2 | 6 | 627 | ||
GenBank mRNA | AK014395.2 | - | 6 | 628 | ||
AK028780.1 | - | 5 | 501 | |||
AK158978.1 | - | 7 | 958 | |||
AK169711.1 | - | 6 | 616 | |||
BC006732.1 | - | 6 | 667 | 3963612 | ||
U16297.1 | - | 6 | 647 |