Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301284.1 | NP_001288213.1 | 5 | 476 | ||
NM_007806.3 | NP_031832.2 | 6 | 560 | |||
GenBank mRNA | AK018713.1 | - | 5 | 445 | ||
AK021200.1 | - | 6 | 548 | |||
AK152414.1 | - | 6 | 551 | |||
AK171748.1 | - | 6 | 529 | |||
BC026791.1 | - | 6 | 511 | 3980538 | ||
BC060635.1 | - | 5 | 4562 | |||
CF738398.1 | - | 6 | 543 | |||
EU791539.1 | - | 6 | 490 | |||
M31775.1 | - | 6 | 483 |