Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007807.5 | NP_031833.3 | 7 | 796 | ||
GenBank mRNA | AK033570.1 | - | 7 | 740 | ||
AK053920.1 | - | 7 | 740 | |||
AK152257.1 | - | 7 | 767 | |||
AK153154.1 | - | 7 | 767 | |||
AK153259.1 | - | 7 | 767 | |||
AK154126.1 | - | 7 | 796 | |||
AK160031.1 | - | 2 | 198 | |||
AK171526.1 | - | 7 | 782 | |||
BC042838.1 | - | 7 | 763 | |||
BC071229.1 | - | 7 | 751 | 4979804 | ||
FJ168469.1 | - | 7 | 785 | |||
U43384.1 | - | 7 | 722 |