Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136059.2 | NP_001129531.1 | 2 | 480 | ||
NM_009992.4 | NP_034122.1 | 2 | 559 | |||
GenBank mRNA | AK005000.1 | - | 2 | 560 | ||
BC125440.1 | - | 2 | 479 | 40129955 | ||
BC125444.1 | - | 2 | 479 | 40129959 | ||
K02588.1 | - | 2 | 479 | |||
Y00071.1 | - | 2 | 480 |