Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009993.3 | NP_034123.1 | 3 | 931 | ||
GenBank mRNA | BC018298.1 | - | 3 | 911 | 4196676 | |
BC054827.1 | - | 3 | 924 | 5054733 | ||
K02589.1 | - | 3 | 932 | |||
X00479.1 | - | 3 | 932 | |||
X04283.1 | - | 2 | 895 |