Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009994.1 | NP_034124.1 | 3 | 1837 | ||
GenBank mRNA | AK017127.1 | - | 2 | 825 | ||
AK041078.1 | - | 2 | 773 | |||
AK137461.1 | - | 3 | 1756 | |||
BC050063.1 | - | 3 | 1837 | 30007248 | ||
U03283.1 | - | 3 | 1837 | |||
X78445.1 | - | 3 | 1660 |