Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp21a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009995.2 | NP_034125.2 | 7 | 778 | ||
GenBank mRNA | AK146565.1 | - | 7 | 915 | ||
BC127167.1 | - | 7 | 751 | 40105690 |