Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009996.3 | NP_034126.1 | 9, 10 | 1606 | ||
GenBank mRNA | AK143984.1 | - | 9, 10 | 1614 | ||
AK159109.1 | - | 9, 10 | 1614 | |||
AK159481.1 | - | 9, 10 | 1614 | |||
AK159527.1 | - | 9, 10 | 1614 | |||
AK159852.1 | - | 9, 10 | 1614 | |||
AK159954.1 | - | 9, 10 | 1614 | |||
AK159976.1 | - | 9, 10 | 1614 | |||
AK164697.1 | - | 9, 10 | 1614 | |||
BC109139.1 | - | 9, 10 | 1598 | 40039500 | ||
BC109140.1 | - | 9, 10 | 1598 | 40039502 | ||
D49438.1 | - | 9, 10 | 1544 | |||
D89669.1 | - | 9, 10 | 1606 |