Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007812.4 | NP_031838.2 | 2 | 230 | ||
GenBank mRNA | AK132617.1 | - | 2 | 229 | ||
AK149439.1 | - | 2 | 231 | |||
BC046605.1 | - | 2 | 204 | |||
BC058743.1 | - | 2 | 202 | |||
BC138796.1 | - | 2 | 226 | 8861818 | ||
BC138798.1 | - | 2 | 226 | 8861820 | ||
X89864.1 | - | 2 | 208 |
Gene Symbol: Cyp2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009997.2 | NP_034127.2 | 2 | 231 | ||
GenBank mRNA | BC010761.1 | - | 2 | 222 | 4239813 | |
BC011233.1 | - | 207 | 4190317 | |||
BC063778.1 | - | 2 | 254 | 5044482 |