Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007812.4 | NP_031838.2 | 9 | 1410 | ||
GenBank mRNA | AK132617.1 | - | 9 | 1409 | ||
AK149439.1 | - | 9 | 1411 | |||
BC046605.1 | - | 9 | 1384 | |||
BC058743.1 | - | 9 | 1382 | |||
BC138796.1 | - | 9 | 1406 | 8861818 | ||
BC138798.1 | - | 9 | 1406 | 8861820 | ||
X89864.1 | - | 9 | 1388 |
Gene Symbol: Cyp2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009997.2 | NP_034127.2 | 9 | 1411 | ||
GenBank mRNA | BC010761.1 | - | 9 | 1402 | 4239813 | |
BC011233.1 | - | 1387 | 4190317 | |||
BC063778.1 | - | 9 | 1434 | 5044482 |