Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039555.2 | NP_001034644.2 | 4 | 655 | ||
GenBank mRNA | AK136413.1 | - | 3 | 285 | ||
BC019468.1 | - | 4 | 608 | 4160492 |
Gene Symbol: Cyp2c40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010004.2 | NP_034134.2 | 4 | 687 | ||
GenBank mRNA | AF047727.1 | - | 4 | 601 | ||
BC139471.1 | - | 4 | 687 | 8862493 | ||
BC139472.1 | - | 4 | 687 | 8862494 |
Gene Symbol: Cyp2c67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024719.2 | NP_001019890.1 | 4 | 607 | ||
GenBank mRNA | AK149389.1 | - | 4 | 618 | ||
BC092260.1 | - | 4 | 607 | 5132591 | ||
BC106142.1 | - | 4 | 607 | 5051946 |
Gene Symbol: Cyp2c69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104525.1 | NP_001097995.1 | 4 | 657 | ||
GenBank mRNA | BC145630.1 | - | 4 | 616 |