Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010006.2 | NP_034136.2 | 7 | 1091 | ||
GenBank mRNA | AK143963.1 | - | 7 | 1091 | ||
BC010593.1 | - | 7 | 1066 | 3964178 | ||
BC061505.1 | - | 1 | 224 | |||
BC094015.1 | - | 7 | 1074 | 5050233 | ||
M23997.1 | - | 6 | 1040 | |||
M23998.1 | - | 7 | 1087 | |||
M27168.1 | - | 7 | 1066 |
Gene Symbol: Cyp2d12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201360.1 | NP_958748.1 | 7 | 1065 | ||
GenBank mRNA | AK078880.1 | - | 7 | 1065 | ||
AK165474.1 | - | 2314 | ||||
BC026417.1 | - | 6 | 1338 | |||
BC089596.1 | - | 3 | 2161 | |||
BC139169.1 | - | 7 | 1049 | 8862191 | ||
BC139170.1 | - | 7 | 1049 | 8862192 | ||
BC145567.1 | - | 6 | 896 | 9054132 |