Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010008.4 | NP_034138.3 | 2 | 499 | ||
GenBank mRNA | AK033614.1 | - | 2 | 502 | ||
AK045421.1 | - | 2 | 489 | |||
AK078893.1 | - | 2 | 325 | |||
AK084025.1 | - | 2 | 509 | |||
AK165663.1 | - | 2 | 344 | |||
AK180579.1 | - | 2 | 188 | |||
AK186838.1 | - | 2 | 188 | |||
AK193212.1 | - | 2 | 188 | |||
AK210777.1 | - | 2 | 285 | |||
BC050832.1 | - | 2 | 493 | 6514145 | ||
U62295.1 | - | 2 | 546 |