Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007818.3 | NP_031844.1 | 5 | 448 | ||
GenBank mRNA | AK004861.2 | - | 1397 | |||
AK149433.1 | - | 5 | 459 | |||
BC010528.1 | - | 5 | 448 | 4194909 | ||
BC094062.1 | - | 5 | 444 | 4162188 | ||
X60452.1 | - | 5 | 444 |