Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007818.3 | NP_031844.1 | 9 | 911 | ||
GenBank mRNA | AK004861.2 | - | 1860 | |||
AK149433.1 | - | 9 | 922 | |||
BC010528.1 | - | 9 | 911 | 4194909 | ||
BC094062.1 | - | 9 | 907 | 4162188 | ||
X60452.1 | - | 9 | 907 |
Gene Symbol: Cyp3a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177380.3 | NP_796354.1 | 9 | 918 | ||
GenBank mRNA | AB039380.1 | - | 9 | 918 | ||
BC125522.1 | - | 9 | 880 | 40130037 |